Assay Details
| Gene Symbol: GRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064760.1 | XP_017920249.1 | 2 - 3 | 493 | 64 | |
| GenBank mRNA | JO648481.2 | - | 2 - 3 | 247 | 64 | |
More Information
Set Membership: |
|

GRAP
Genomic Map