Assay Details
| Gene Symbol: CRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044460.1 | XP_017899949.1 | 3 - 4 | 276 | 99 | |
| GenBank mRNA | KA323032.1 | - | 3 - 4 | 261 | 99 | |
More Information
Set Membership: |
|

CRIP1
Genomic Map