Assay Details
| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041602.1 | XP_017897091.1 | 2 - 3 | 694 | 80 | |
| GenBank mRNA | JO423013.2 | - | 1 - 2 | 589 | 80 | |
| KU161270.1 | - | 4 - 5 | 734 | 80 | ||
More Information
Set Membership: |
|

FFAR4
Genomic Map