Assay Details
| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678741.3 | XP_005678798.2 | 2 - 3 | 260 | 94 | |
| XM_013963020.2 | XP_013818474.2 | 2 - 3 | 400 | 94 | ||
| GenBank mRNA | JO419417.2 | - | 1 - 2 | 407 | 94 | |
| KA316739.1 | - | 2 - 3 | 1838 | 94 | ||
More Information
Set Membership: |
|

NEU2
Genomic Map