Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690034.3 | XP_005690091.1 | 2 - 3 | 193 | 79 | |
| XM_005690035.3 | XP_005690092.1 | 2 - 3 | 234 | 79 | ||
| XM_005690036.3 | XP_005690093.1 | 2 - 3 | 193 | 79 | ||
| XM_013969800.2 | XP_013825254.1 | 2 - 3 | 234 | 79 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map