Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058274.1 | XP_017913763.1 | 2 - 3 | 1409 | 82 | |
| GenBank mRNA | JO414154.2 | - | 1 - 2 | 338 | 82 | |
More Information
Set Membership: |
|

ERICH5
Genomic Map