Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062173.1 | XP_017917662.1 | 2 - 3 | 1154 | 73 | |
| GenBank mRNA | JO425834.1 | - | 2 - 3 | 1253 | 73 | |
| JT678434.1 | - | 2 - 3 | 1185 | 73 | ||
More Information
Set Membership: |
|

PSKH1
Genomic Map