Assay Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064758.1 | XP_017920247.1 | 1 - 2 | 603 | 67 | |
| GenBank mRNA | JO584058.1 | - | 1 - 2 | 204 | 67 | |
More Information
Set Membership: |
|

FAM83G
Genomic Map