Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677116.3 | XP_005677173.1 | 1 - 2 | 186 | 59 | |
| GenBank mRNA | JO572628.2 | - | 4 - 5 | 405 | 59 | |
More Information
Set Membership: |
|

RGS4
Genomic Map