Assay Details
| Gene Symbol: PIGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JT669132.1 | - | 1 | 1191 | 93 | |
| KA334536.1 | - | 1 | 1315 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PIGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JT669132.1 | - | 1 | 1191 | 93 | |
| KA334536.1 | - | 1 | 1315 | 93 | ||
Set Membership: |
|