Assay Details
| Gene Symbol: VPS72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677621.3 | XP_005677678.1 | 5 - 6 | 800 | 61 | |
| GenBank mRNA | JO637741.2 | - | 5 - 6 | 795 | 61 | |
More Information
Set Membership: |
|

VPS72
Genomic Map