Assay Details
| Gene Symbol: NDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681260.3 | XP_005681317.2 | 3 - 4 | 726 | 81 | |
| GenBank mRNA | JO506950.2 | - | 1 - 2 | 412 | 81 | |
More Information
Set Membership: |
|

NDNF
Genomic Map