Assay Details
| Gene Symbol: IL11RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052397.1 | XP_017907886.1 | 3 - 4 | 220 | 55 | |
| XM_018052398.1 | XP_017907887.1 | 3 - 4 | 256 | 55 | ||
| XM_018052399.1 | XP_017907888.1 | 3 - 4 | 277 | 55 | ||
| XM_018052400.1 | XP_017907889.1 | 3 - 4 | 639 | 55 | ||
| XM_018052401.1 | XP_017907890.1 | 3 - 4 | 496 | 55 | ||
More Information
Set Membership: |
|

IL11RA
Genomic Map