Assay Details
| Gene Symbol: MRPL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060525.1 | XP_017916014.1 | 3 - 4 | 362 | 84 | |
| GenBank mRNA | JO599024.2 | - | 1 - 2 | 460 | 84 | |
| KA336845.1 | - | 1 - 2 | 397 | 84 | ||
More Information
Set Membership: |
|

MRPL20
Genomic Map