Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061574.1 | XP_017917063.1 | 6 - 7 | 1702 | 57 | |
| XM_018061575.1 | XP_017917064.1 | 6 - 7 | 1480 | 57 | ||
| XM_018061576.1 | XP_017917065.1 | 6 - 7 | 1448 | 57 | ||
| XM_018061578.1 | XP_017917067.1 | 6 - 7 | 1492 | 57 | ||
More Information
Set Membership: |
|

SMAD1
Genomic Map