Assay Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048111.1 | XP_017903600.1 | 2 - 3 | 868 | 93 | |
| GenBank mRNA | JO634672.2 | - | 2 - 3 | 739 | 93 | |
More Information
Set Membership: |
|

GDF11
Genomic Map