Assay Details
| Gene Symbol: ITPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688203.3 | XP_005688260.1 | 5 - 6 | 411 | 67 | |
| GenBank mRNA | JO355614.2 | - | 5 - 6 | 323 | 67 | |
| JO442732.2 | - | 3 - 4 | 629 | 67 | ||
More Information
Set Membership: |
|

ITPA
Genomic Map