Assay Details
Gene Symbol: GPX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695962.3 | XP_005696019.2 | 1 - 2 | 356 | 58 | |
GenBank mRNA | JO443233.2 | - | 1 - 2 | 386 | 58 | |
JO633492.2 | - | 1 - 2 | 432 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
