Assay Details
| Gene Symbol: MRPL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040330.1 | XP_017895819.1 | 5 - 6 | 882 | 70 | |
| XM_018040331.1 | XP_017895820.1 | 4 - 5 | 729 | 70 | ||
More Information
Set Membership: |
|

MRPL28
Genomic Map