Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678094.3 | XP_005678151.1 | 1 - 2 | 442 | 92 | |
| XM_005678095.2 | XP_005678152.1 | 1 - 2 | 414 | 92 | ||
| GenBank mRNA | KA325409.1 | - | 1 - 2 | 76 | 92 | |
More Information
Set Membership: |
|

TMED5
Genomic Map