Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678562.3 | XP_005678619.2 | 1 - 2 | 720 | 71 | |
| XM_018043100.1 | XP_017898589.1 | 1 - 2 | 720 | 71 | ||
| GenBank mRNA | JO609901.2 | - | 1 - 2 | 113 | 71 | |
| KA353371.1 | - | 1 - 2 | 394 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018043100.1 transcripts.
Set Membership: |
|

DPH2
Genomic Map