Assay Details
| Gene Symbol: RRAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683637.3 | XP_005683694.1 | 1 | 1110 | 108 | |
| GenBank mRNA | JO371653.2 | - | 1 | 908 | 108 | |
| KA315605.1 | - | 1 | 930 | 108 | ||
More Information
Set Membership: |
|

RRAGA
Genomic Map