Assay Details
| Gene Symbol: MALRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687912.3 | XP_005687969.3 | 1 - 2 | 180 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MALRD1| Gene Symbol: MALRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687912.3 | XP_005687969.3 | 1 - 2 | 180 | 64 | |
Set Membership: |
|