Assay Details
| Gene Symbol: NDUFB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700819.2 | XP_005700876.1 | 1 - 2 | 487 | 60 | |
| GenBank mRNA | JO444039.2 | - | 1 - 2 | 514 | 60 | |
More Information
Set Membership: |
|

NDUFB11
Genomic Map