Assay Details
| Gene Symbol: TTLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048867.1 | XP_017904356.1 | 4 - 5 | 542 | 63 | |
| GenBank mRNA | JO499982.2 | - | 4 - 5 | 483 | 63 | |
| JO630785.2 | - | 7 - 8 | 1094 | 63 | ||
More Information
Set Membership: |
|

TTLL1
Genomic Map