Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051448.1 | XP_017906937.1 | 7 - 8 | 1015 | 64 | |
| XM_018051449.1 | XP_017906938.1 | 6 - 7 | 851 | 64 | ||
| GenBank mRNA | JT763257.1 | - | 7 - 8 | 904 | 64 | |
More Information
Set Membership: |
|

TMEM59L
Genomic Map