Assay Details
| Gene Symbol: P4HTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066757.1 | XP_017922246.1 | 2 - 3 | 445 | 74 | |
| GenBank mRNA | JO634674.2 | - | 7 - 8 | 1321 | 74 | |
More Information
Set Membership: |
|

P4HTM
Genomic Map