Assay Details
| Gene Symbol: PCBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042610.1 | XP_017898099.1 | 3 - 4 | 321 | 92 | |
| GenBank mRNA | JO535714.2 | - | 1 - 2 | 550 | 92 | |
More Information
Set Membership: |
|

PCBD1
Genomic Map