Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677414.2 | XP_005677471.1 | 3 - 4 | 1052 | 62 | |
| XM_005677415.3 | XP_005677472.1 | 3 - 4 | 1294 | 62 | ||
| XM_005677416.3 | XP_005677473.1 | 3 - 4 | 1015 | 62 | ||
| XM_005677417.3 | XP_005677474.1 | 3 - 4 | 1064 | 62 | ||
| GenBank mRNA | JO410526.2 | - | 3 - 4 | 1045 | 62 | |
| JO604853.2 | - | 1 - 2 | 201 | 62 | ||
More Information
Set Membership: |
|

FLAD1
Genomic Map