Assay Details
| Gene Symbol: SCFD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681611.3 | XP_005681668.2 | 3 - 4 | 1256 | 61 | |
| GenBank mRNA | JO608287.2 | - | 3 - 4 | 788 | 61 | |
More Information
Set Membership: |
|

SCFD2
Genomic Map