Assay Details
| Gene Symbol: RPL10L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685551.2 | XP_005685608.1 | 1 | 617 | 118 | |
| XM_005685552.2 | XP_005685609.1 | 2 | 495 | 118 | ||
More Information
Set Membership: |
|

RPL10L
Genomic Map