Assay Details
| Gene Symbol: PSMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057594.1 | XP_017913083.1 | 3 - 4 | 491 | 67 | |
| XM_018057595.1 | XP_017913084.1 | 3 - 4 | 491 | 67 | ||
| GenBank mRNA | JO631709.2 | - | 3 - 4 | 501 | 67 | |
| KA328464.1 | - | 3 - 4 | 455 | 67 | ||
More Information
Set Membership: |
|

PSMA7
Genomic Map