Assay Details
| Gene Symbol: LOC102172084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059551.1 | XP_017915040.1 | 35 - 36 | 6000 | 108 | |
| XM_018059552.1 | XP_017915041.1 | 35 - 36 | 5996 | 108 | ||
| XM_018059553.1 | XP_017915042.1 | 35 - 36 | 5997 | 108 | ||
| XM_018059554.1 | XP_017915043.1 | 34 - 35 | 5937 | 108 | ||
| XM_018059555.1 | XP_017915044.1 | 35 - 36 | 5877 | 108 | ||
| XM_018059556.1 | XP_017915045.1 | 33 - 34 | 5811 | 108 | ||
| XM_018059557.1 | XP_017915046.1 | 31 - 32 | 5430 | 108 | ||
| XM_018059558.1 | XP_017915047.1 | 30 - 31 | 5367 | 108 | ||
| XM_018059559.1 | XP_017915048.1 | 30 - 31 | 5322 | 108 | ||
| XM_018059560.1 | XP_017915049.1 | 29 - 30 | 5258 | 108 | ||
More Information
Set Membership: |
|

LOC102172084
Genomic Map