Assay Details
| Gene Symbol: SMARCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060907.1 | XP_017916396.1 | 8 - 9 | 1343 | 66 | |
| XM_018060908.1 | XP_017916397.1 | 8 - 9 | 1316 | 66 | ||
| GenBank mRNA | JO610910.2 | - | 8 - 9 | 1280 | 66 | |
More Information
Set Membership: |
|

SMARCB1
Genomic Map