Assay Details
| Gene Symbol: LCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697619.2 | XP_005697676.1 | 1 - 2 | 258 | 64 | |
| GenBank mRNA | JO645592.2 | - | 10 - 11 | 1140 | 64 | |
More Information
Set Membership: |
|

LCMT1
Genomic Map