Assay Details
| Gene Symbol: SAMM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048880.1 | XP_017904369.1 | 7 - 8 | 842 | 60 | |
| GenBank mRNA | JO556196.2 | - | 7 - 8 | 767 | 60 | |
| JO640060.2 | - | 1 - 2 | 81 | 60 | ||
More Information
Set Membership: |
|

SAMM50
Genomic Map