Assay Details
| Gene Symbol: CRELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045509.1 | XP_017900998.1 | 8 - 9 | 1011 | 58 | |
| GenBank mRNA | JO581043.1 | - | 2 - 3 | 474 | 58 | |
More Information
Set Membership: |
|

CRELD2
Genomic Map