Assay Details
| Gene Symbol: PRDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053115.1 | XP_017908604.1 | 5 - 6 | 2302 | 66 | |
| XM_018053116.1 | XP_017908605.1 | 5 - 6 | 2020 | 66 | ||
| XM_018053117.1 | XP_017908606.1 | 3 - 4 | 2401 | 66 | ||
More Information
Set Membership: |
|

PRDM1
Genomic Map