Assay Details
| Gene Symbol: CCNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066176.1 | XP_017921665.1 | 7 - 8 | 1087 | 66 | |
| GenBank mRNA | JO600588.2 | - | 4 - 5 | 714 | 66 | |
| KA320195.1 | - | 7 - 8 | 900 | 66 | ||
More Information
Set Membership: |
|

CCNK
Genomic Map