Assay Details
| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066630.1 | XP_017922119.1 | 4 - 5 | 814 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A13| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066630.1 | XP_017922119.1 | 4 - 5 | 814 | 62 | |
Set Membership: |
|