Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680822.3 | XP_005680879.2 | 3 - 4 | 701 | 68 | |
| GenBank mRNA | JO607296.2 | - | 1 - 2 | 160 | 68 | |
| JT689542.1 | - | 3 - 4 | 623 | 68 | ||
More Information
Set Membership: |
|

MANSC1
Genomic Map