Assay Details
| Gene Symbol: GTF2H5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684941.3 | XP_005684998.1 | 1 - 2 | 80 | 60 | |
| GenBank mRNA | KA342928.1 | - | 2 - 3 | 456 | 60 | |
More Information
Set Membership: |
|

GTF2H5
Genomic Map