Assay Details
| Gene Symbol: SLC5A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967806.2 | XP_013823260.1 | 12 - 13 | 1633 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A6| Gene Symbol: SLC5A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967806.2 | XP_013823260.1 | 12 - 13 | 1633 | 60 | |
Set Membership: |
|