Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689055.3 | XP_005689112.2 | 1 - 2 | 210 | 66 | |
| GenBank mRNA | JO434305.2 | - | 1 - 2 | 208 | 66 | |
More Information
Set Membership: |
|

LACTB2
Genomic Map