Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689699.3 | XP_005689756.1 | 2 - 3 | 283 | 93 | |
| XM_013969639.2 | XP_013825093.1 | 2 - 3 | 599 | 93 | ||
| XM_018059529.1 | XP_017915018.1 | 2 - 3 | 285 | 93 | ||
| XM_018059530.1 | XP_017915019.1 | 3 - 4 | 366 | 93 | ||
| GenBank mRNA | JO423661.2 | - | 2 - 3 | 940 | 93 | |
More Information
Set Membership: |
|

ADM
Genomic Map