Assay Details
| Gene Symbol: OPCML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975837.2 | XP_013831291.1 | 7 - 8 | 1348 | 64 | |
| XM_018043456.1 | XP_017898945.1 | 7 - 8 | 1345 | 64 | ||
More Information
Set Membership: |
|

OPCML
Genomic Map