Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699636.3 | XP_005699693.2 | 5 - 6 | 911 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM45B| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699636.3 | XP_005699693.2 | 5 - 6 | 911 | 59 | |
Set Membership: |
|