Assay Details
| Gene Symbol: FGF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043636.1 | XP_017899125.1 | 2 - 3 | 703 | 96 | |
| GenBank mRNA | JT762023.1 | - | 2 - 3 | 331 | 96 | |
More Information
Set Membership: |
|

FGF16
Genomic Map