Assay Details
| Gene Symbol: GPRC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063873.1 | XP_017919362.1 | 4 - 5 | 1450 | 65 | |
| XM_018063875.1 | XP_017919364.1 | 4 - 5 | 2506 | 65 | ||
| XM_018063876.1 | XP_017919365.1 | 5 - 6 | 2379 | 65 | ||
| XM_018063877.1 | XP_017919366.1 | 4 - 5 | 2119 | 65 | ||
| XM_018063878.1 | XP_017919367.1 | 4 - 5 | 1621 | 65 | ||
More Information
Set Membership: |
|

GPRC5C
Genomic Map