Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962541.2 | XP_013817995.1 | 6 - 7 | 492 | 74 | |
| GenBank mRNA | JO556268.2 | - | 6 - 7 | 520 | 74 | |
| JO564202.2 | - | 6 - 7 | 601 | 74 | ||
More Information
Set Membership: |
|

RAP1A
Genomic Map